3-Bromo-4-formylpyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Bromo-4-pyridinecarboxaldehyde, 3-Bromoisonicotinaldehyde, 3-Bromopyridine-4-carboxaldehyde

39808-0010 39808-0050
TS39808-0010EA 132 USD
TS39808-0010 TS39808-0050
3-Bromo-4-formylpyridine 97%
3-Bromo-4-formylpyridine
Formula: C₆H₄BrNO
MW: 186.01 g/mol
Melting Pt: 80…83 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05864506
CAS Number: 70201-43-3

Order Now

Specification Test Results

Appearance Light yellow to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 79 to 83 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR