2-Bromo-6-methoxypyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
39767-0050
39767-0250
TS39767-0010EA
21.63
USD
TS39767-0010
TS39767-0050
TS39767-0250
2-Bromo-6-methoxypyridine 97%
2-Bromo-6-methoxypyridine
Formula:
C₆H₆BrNO Boiling Pt: 206 °C Flash Pt: 104 °C Storage Temperature: Ambient |
MDL Number:
MFCD00088345 CAS Number: 40473-07-2 UN: 2810 ADR: 6.1,III |
Specification Test Results
Appearance | Clear colorless to golden Liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...