2,2'-Thenil 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2-Di(thiophen-2-yl)ethane-1,2-dione

39363-0010 39363-0050
TS39363-0010EA 48 USD
TS39363-0010 TS39363-0050
2,2'-Thenil 98+%
2,2'-Thenil
Formula: C₁₀H₆O₂S₂
MW: 222.29 g/mol
Melting Pt: 81…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00173678
CAS Number: 7333-07-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow Powder
Infrared spectrum Conforms
Melting point 81 to 84 °C
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR