1,1-Diethoxybutane 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Butyraldehyde diethylacetal

39343-0050 39343-0250 39343-1000
TS39343-0050EA 31 USD
TS39343-0050 TS39343-0250 TS39343-1000
1,1-Diethoxybutane 97%
1,1-Diethoxybutane
Formula: C₈H₁₈O₂
Boiling Pt: 143 °C (760 mmHg)
Density: 0.83 g/cm³
Flash Pt: 30 °C
Storage Temperature: Ambient
CAS Number: 3658-95-5
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3950 to 1.3980 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR