(S)-1-(Pyridin-4-yl)ethanol 99+%, (99+% ee)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-alpha-Methyl-4-pyridinemethanol, (S)-1-(4-Pyridyl)ethanol, (S)-4-(1-Hydroxyethyl)pyridine

39337-0010 39337-2500
TS39337-0010EA 360 USD
TS39337-0010 TS39337-2500
(S)-1-(Pyridin-4-yl)ethanol 99+%, (99+% ee)
(S)-1-(Pyridin-4-yl)ethanol
Formula: C₇H₉NO
MW: 123.155 g/mol
Melting Pt: 67…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077866
CAS Number: 54656-96-1

Order Now

Specification Test Results

Appearance White to cream Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 67 to 70 °C
GC ≥99.0 %
Enantiomeric excess ≥99.0 %
Specific optical rotation -56° to -61° (20 °C, 589 nm) (c=1, chloroform)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR