2'-Iodoacetophenone 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(2-Iodophenyl)ethan-1-one, 1-(2-Iodophenyl)ethanone, o-Iodoacetophenone

39313-0010 39313-0050
TS39313-0010EA 31 USD
TS39313-0010 TS39313-0050
2'-Iodoacetophenone 99+%
2'-Iodoacetophenone
Formula: C₈H₇IO
MW: 246.05 g/mol
Boiling Pt: 139…140 °C (12 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00094998
CAS Number: 2142-70-3
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6170 to 1.6200 (20 °C, 589 nm)
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR