Methyl 6-bromohexanoate 96+%

Supplier: Thermo Scientific Chemicals
Warning

39269-0050 39269-0250 39269-1000
TS39269-0050EA 43 USD
TS39269-0050 TS39269-0250 TS39269-1000
Methyl 6-bromohexanoate 96+%
Methyl 6-bromohexanoate
Formula: C₇H₁₃BrO₂
MW: 209.084 g/mol
Boiling Pt: 150 °C (50 mmHg)
Density: 1.316 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00187528
CAS Number: 14273-90-6

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4630 to 1.4650 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR