3,7-Dimethyl-1-octanol 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
39262-0500
39262-2500
TS39262-0500EA
37
USD
TS39262-0500
TS39262-2500
3,7-Dimethyl-1-octanol 95%
3,7-Dimethyl-1-octanol
Formula:
C₁₀H₂₂O Boiling Pt: 118 °C (15 mmHg) Density: 0.828 g/cm³ Flash Pt: 97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002936 CAS Number: 106-21-8 UN: 3082 ADR: 9,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4340 to 1.4360 (20 °C, 589 nm) |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...