4,4'-Biphenyldicarboxaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: [1,1'-Biphenyl]-4,4'-dicarbaldehyde, 4,4'-Diformylbiphenyl

39113-0010 39113-0050
TS39113-0010EA 109 USD
TS39113-0010 TS39113-0050
4,4'-Biphenyldicarboxaldehyde 97%
4,4'-Biphenyldicarboxaldehyde
Formula: C₁₄H₁₀O₂
MW: 210.23 g/mol
Melting Pt: 144…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00016714
CAS Number: 66-98-8

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 144 to 147 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR