4'-Ethynylacetophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(4-Ethynyl-phenyl)-ethanone, 1-(4-Ethynylphenyl)ethanone, 1 - (4 - Ethynyl - phenyl) - ethanone, Ethanone,1-(4-ethynylphenyl)-

39058-0010 39058-0050
TS39058-0010EA 167 USD
TS39058-0010 TS39058-0050
4'-Ethynylacetophenone 98%
4'-Ethynylacetophenone
Formula: C₁₀H₈O
MW: 144.173 g/mol
Melting Pt: 67…70 °C
Storage Temperature: Ambient
MDL Number: MFCD04974025
CAS Number: 42472-69-5

Order Now

Specification Test Results

Appearance Yellow to brown Powder
Infrared spectrum Conforms
Melting point 67 to 70 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR