2-Nitro-1-phenylpropene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
trans-β-Methyl-β-nitrostyrene
38651-0010
38651-0050
38651-0250
TS38651-0010EA
23
USD
TS38651-0010
TS38651-0050
TS38651-0250
2-Nitro-1-phenylpropene 99%
2-Nitro-1-phenylpropene
Formula:
C₉H₉NO₂ Melting Pt: 64…67 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014720 CAS Number: 705-60-2 |
Specification Test Results
Appearance | Yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 64 to 67 °C |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...