Methyl pelargonate 95%

Supplier: Thermo Scientific Chemicals

38556-0050 38556-0250 38556-1000
TS38556-0050EA 55 USD
TS38556-0050 TS38556-0250 TS38556-1000
Methyl pelargonate 95%
Methyl pelargonate
Formula: C₁₀H₂₀O₂
Boiling Pt: 213 °C (760 mmHg)
Melting Pt: –30 °C
Density: 0.874 g/cm³
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00009569
CAS Number: 1731-84-6

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4210 to 1.4230 (20 °C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR