10-Bromo-1-decanol 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 10-Bromodecanol

38459-0010 38459-0050 38459-0250
TS38459-0010EA 33 USD
TS38459-0010 TS38459-0050 TS38459-0250
10-Bromo-1-decanol 95%
10-Bromo-1-decanol
Formula: HO(CH₂)₉CH₂Br
MW: 237.18 g/mol
Boiling Pt: 165…169 °C (10 mmHg)
Density: 1.19 g/cm³
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00041681
CAS Number: 53463-68-6

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4780 to 1.4800 (20 °C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR