2-Methyl-1H-indole-3-carbaldehyde 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methylindole-3-carboxaldehyde, 3-Formyl-2-methylindole

37972-0250 37972-1000
TS37972-0250EA 156 USD
TS37972-0250 TS37972-1000
2-Methyl-1H-indole-3-carbaldehyde 97+%
2-Methyl-1H-indole-3-carbaldehyde
Formula: C₁₀H₉NO
Melting Pt: 204…205 °C
Storage Temperature: Ambient
MDL Number: MFCD00012077
CAS Number: 5416-80-8

Order Now

Specification Test Results

Appearance Brown Powder
Infrared spectrum Conforms
Melting point 199 to 202 °C
HPLC ≥97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR