(±)-3,7-Dimethyl-3-octanol 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tetrahydrolinalool, 3,7-Dimethyloctan-3-ol, 3,7-Dimethyl-3-octanol

37906-1000 37906-5000
TS37906-1000EA 45 USD
TS37906-1000 TS37906-5000
(±)-3,7-Dimethyl-3-octanol 97+%
(±)-3,7-Dimethyl-3-octanol
Formula: C₁₀H₂₂O
MW: 158.28 g/mol
Boiling Pt: 193 °C (760 mmHg)
Density: 0.828 g/cm³
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00004482
CAS Number: 78-69-3
EINECS: 201-133-9

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4310 to 1.4350 (20 °C, 589 nm)
GC ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR