Thiazole-2-carbaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Thiazolecarboxaldehyde, Thiazole-2-carboxaldehyde

37768-0010 37768-0050
TS37768-0010EA 73 USD
TS37768-0010 TS37768-0050
Thiazole-2-carbaldehyde 98%
Thiazole-2-carbaldehyde
Formula: C₄H₃NOS
MW: 113.13 g/mol
Boiling Pt: 76…78 °C (24 mmHg)
Density: 1.288 g/cm³
Flash Pt: 68 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00142924
CAS Number: 10200-59-6

Order Now

Specification Test Results

Appearance Clear yellow to orange-red to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5730 to 1.5760 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR