5-Bromo-2-methoxypyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

37688-0050 37688-0250
TS37688-0050EA 47 USD
TS37688-0050 TS37688-0250
5-Bromo-2-methoxypyridine 97%
5-Bromo-2-methoxypyridine
Formula: C₆H₆BrNO
MW: 188.02 g/mol
Boiling Pt: 80 °C (12 mmHg)
Flash Pt: 96 °C
MDL Number: MFCD01318952
CAS Number: 13472-85-0

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5530 to 1.5570 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR