Nerol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: cis-3,7-Dimethyl-2,6-octadien-1-ol

37654-0250 37654-1000 37654-5000
TS37654-0250EA 34 USD
TS37654-0250 TS37654-1000 TS37654-5000
Nerol 97%
Nerol
Formula: C₁₀H₁₈O
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00063204
CAS Number: 106-25-2
EINECS: 203-378-7

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4720 to 1.4790 (20 °C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR