1-Bromo-2-(2-methoxyethoxy)ethane 90% stabilised

Supplier: Thermo Scientific Chemicals

Synonyms: 1-Bromo-3,6-dioxaheptane, 2-(2-Methoxyethoxy)ethyl bromide, m-PEG2-Br

Stabilization: Stabilised with sodium carbonate (Na₂CO₃)

37098-0100 37098-0500
TS37098-0100EA 108 USD
TS37098-0100 TS37098-0500
1-Bromo-2-(2-methoxyethoxy)ethane 90% stabilised
1-Bromo-2-(2-methoxyethoxy)ethane
Formula: BrH₂CCH₂OCH₂CH₂OCH₃
Boiling Pt: 172 °C (1013 hPa)
Melting Pt: –23 °C
Density: 1.347 g/cm³
Flash Pt: 73 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000238
CAS Number: 54149-17-6
EINECS: 259-000-6

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4540 to 1.4600 (20 °C, 589 nm)
GC ≥88 %
Stabilizer 0.1 % Sodium carbonate


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR