3-Methyl-2-butenal 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3,3-Dimethylacrolein, 3-Methylbut-2-enal, Senecialdehyde, 3-Methylcrotonaldehyde

37018-0050 37018-0250 37018-1000
TS37018-0050EA 74 USD
TS37018-0050 TS37018-0250 TS37018-1000
3-Methyl-2-butenal 97%
3-Methyl-2-butenal
Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 132…133 °C
Density: 0.88 g/cm³
Flash Pt: 33 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010291
CAS Number: 107-86-8
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4600 to 1.4640 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR