Stearyl stearate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Stearic acid stearyl ester, Octadecyl stearate, Octadecyl octadecanoate

36811-0010 36811-0050
TS36811-0010EA 164 USD
TS36811-0010 TS36811-0050
Stearyl stearate 98%
Stearyl stearate
Formula: C₃₆H₇₂O₂
MW: 536.97 g/mol
Boiling Pt: ∼175 °C (1013 hPa)
Melting Pt: 64
Storage Temperature: Ambient
MDL Number: MFCD00056225
CAS Number: 2778-96-3
EINECS: 220-476-5

Order Now

Specification Test Results

Appearance White Powder or platelets
Infrared spectrum Conforms
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR