α-Linolenic acid methyl ester 99%

Supplier: Thermo Scientific Chemicals

36810-0010 36810-0050
TS36810-0010EA 91 USD
TS36810-0010 TS36810-0050
α-Linolenic acid methyl ester 99%
α-Linolenic acid methyl ester
Formula: C₁₉H₃₂O₂
Boiling Pt: 182 °C (3 mmHg)
Melting Pt: –57 °C
Density: 0.895 g/cm³
Flash Pt: >110 °C
Storage Temperature: Freezer
MDL Number: MFCD00135851
CAS Number: 301-00-8
EINECS: 206-102-3

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4700 to 1.4720 (20 °C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR