Methyl-3-formylbenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

36805-0010 36805-0050 36805-0250
TS36805-0010EA 70 USD
TS36805-0010 TS36805-0050 TS36805-0250
Methyl-3-formylbenzoate 98%
Methyl-3-formylbenzoate
Formula: C₉H₈O₃
Melting Pt: 48…55 °C
Storage Temperature: Ambient
MDL Number: MFCD00189379
CAS Number: 52178-50-4

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 48 to 55 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR