8-Bromocaprylic acid 95%

Supplier: Thermo Scientific Chemicals
Danger

36766-0100 36766-0500
TS36766-0100EA 125 USD
TS36766-0100 TS36766-0500
8-Bromocaprylic acid 95%
8-Bromocaprylic acid
Formula: BrH₂C(CH₂)₆COOH
Boiling Pt: 147…150 °C (2 mmHg)
Melting Pt: 36…41 °C
Flash Pt: >110 °C
MDL Number: MFCD00004430
CAS Number: 17696-11-6
UN: 3261

Order Now

Specification Test Results

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point 36 to 41 °C
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR