2-Bromoethyl ethyl ether 90%
Supplier: Thermo Scientific Chemicals
|
Danger
|
36740-0250
36740-1000
TS36740-0250EA
67
USD
TS36740-0250
TS36740-1000
TS36740-5000
2-Bromoethyl ethyl ether 90%
2-Bromoethyl ethyl ether
Formula:
C₄H₉BrO MW: 153.02 g/mol Boiling Pt: 125…127 °C Density: 1.357 g/cm³ Flash Pt: 21 °C Storage Temperature: Freezer |
MDL Number:
MFCD00000237 CAS Number: 592-55-2 EINECS: 209-763-6 UN: 2340 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4435 to 1.4515 (20 °C, 589 nm) |
GC | ≥88 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...