Methyl-4-bromobutyrate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 4-bromobutanoate

36583-0100 36583-0500
TS36583-0100EA 52 USD
TS36583-0100 TS36583-0500
Methyl-4-bromobutyrate 98+%
Methyl-4-bromobutyrate
Formula: C₅H₉BrO₂
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00041482
CAS Number: 4897-84-1
EINECS: 225-523-3

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4610 to 1.4630 (20 °C, 589 nm)
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR