(1R,2S,3R,5R)-3-amino-5-(hydroxymethyl)cyclopentane-1,2-diol hydrochloride 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(1R,2S,3R,4R)-2,3-Dihydroxy-4-(hydroxymethyl)-1-aminocyclopentane hydrochloride, (1R,2S,3R,5R)-3-Amino-5-(hydroxymethyl)cyclopentane-1,2-diol hydrochloride
36252-0010
36252-0050
36252-2500
TS36252-0010EA
255
USD
TS36252-0010
TS36252-0050
TS36252-2500
(1R,2S,3R,5R)-3-amino-5-(hydroxymethyl)cyclopentane-1,2-diol hydrochloride 95%
(1R,2S,3R,5R)-3-amino-5-(hydroxymethyl)cyclopentane-1,2-diol hydrochloride
Formula:
C₆H₁₄ClNO₃ MW: 183.632 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06799068 CAS Number: 79200-57-0 |
Specification Test Results
Appearance | White to off-white Crystalline powder and/or chunks |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
Specific optical rotation | -16° to -20° (20°C, 589 nm) (c=1, methanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...