2-Methyl-2-pentenal 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Methylpent-2-enal

35996-0250 35996-1000
TS35996-0250EA 43 USD
TS35996-0250 TS35996-1000
2-Methyl-2-pentenal 97%
2-Methyl-2-pentenal
Formula: C₆H₁₀O
MW: 98.14 g/mol
Boiling Pt: 134…136 °C (760 mmHg)
Density: 0.86 g/cm³
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00006978
CAS Number: 623-36-9
EINECS: 210-789-5
UN: 1988
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4470 to 1.4550 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR