2,5-Dichlorobenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Danger

35994-0010 35994-0050
TS35994-0010EA 33 USD
TS35994-0010 TS35994-0050
2,5-Dichlorobenzaldehyde 98%
2,5-Dichlorobenzaldehyde
Formula: C₇H₄Cl₂O
Melting Pt: 54…58 °C
MDL Number: MFCD00156140
CAS Number: 6361-23-5
EINECS: 228-832-1
UN: 3261

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 54 to 58 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR