trans-2-Methyl-2-butenal 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trans-2-methyl-2-butenal, Tiglic aldehyde, (E)-2-Methyl-2-butenal

35303-0050 35303-0250
TS35303-0050EA 47 USD
TS35303-0050 TS35303-0250
trans-2-Methyl-2-butenal 98%
trans-2-Methyl-2-butenal
Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 115…117 °C (1013 hPa)
Density: 0.871 g/cm³
Flash Pt: 18 °C
MDL Number: MFCD00006977
CAS Number: 497-03-0
EINECS: 207-833-0
UN: 1989
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR