2-Naphthalenemethanol 98%

Supplier: Thermo Scientific Chemicals

35115-0010 35115-0050 35115-0250
TS35115-0010EA 34 USD
TS35115-0010 TS35115-0050 TS35115-0250
2-Naphthalenemethanol 98%
2-Naphthalenemethanol
Formula: C₁₁H₁₀O
MW: 158.2000 g/mol
Melting Pt: 79…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00004124
CAS Number: 1592-38-7

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 79 to 83 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR