2-(3,4-Dichlorophenyl)ethanol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3,4-Dichlorophenethyl Alcohol
35077-0010
35077-0050
TS35077-0010EA
45
USD
TS35077-0010
TS35077-0050
2-(3,4-Dichlorophenyl)ethanol 97%
2-(3,4-Dichlorophenyl)ethanol
Formula:
C₈H₈Cl₂O MW: 191.05 g/mol Boiling Pt: 165 °C (14 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00800673 CAS Number: 35364-79-5 UN: 2810 ADR: 6.1,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5620 to 1.5680 (20 °C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...