Pentaethylene glycol 98%

Supplier: Thermo Scientific Chemicals
Warning

35073-0050 35073-0250
TS35073-0050EA 96 USD
TS35073-0050 TS35073-0250
Pentaethylene glycol 98%
Pentaethylene glycol
Formula: C₁₀H₂₂O₆
Storage Temperature: Ambient
MDL Number: MFCD00002878
CAS Number: 4792-15-8
EINECS: 225-341-4

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4610 to 1.4630 (20 °C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR