3-Bromo-1-propanol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Bromopropan-1-ol, 1-Bromo-3-hydroxypropane

34970-0050 34970-0250 34970-1000
TS34970-0050EA 50 USD
TS34970-0050 TS34970-0250 TS34970-1000
3-Bromo-1-propanol 97%
3-Bromo-1-propanol
Formula: BrH₂CCH₂CH₂OH
Boiling Pt: 62 °C (5 mmHg)
Density: 1.6 g/cm³
Flash Pt: 65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002942
CAS Number: 627-18-9
EINECS: 210-986-6

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4860 to 1.4900 (20 °C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR