Dodecyl methacrylate 97% stabilised

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dodecyl 2-methylacrylate, Lauryl methacrylate

34771-0025 34771-1000 34771-5000
TS34771-0025EA 284 USD
TS34771-0025 TS34771-1000 TS34771-5000
Dodecyl methacrylate 97% stabilised
Dodecyl methacrylate
Formula: C₁₆H₃₀O₂
MW: 254.41 g/mol
Melting Pt: –23 °C
Density: 0.871 g/cm³
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD00008972
CAS Number: 142-90-5

Order Now

Specification Test Results

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4470 (20 °C, 589 nm)
GC ≥96.0 %
Stabilizer 300 to 500 ppm MEHQ


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR