3-Mercapto-1-hexanol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Mercaptohexan-1-ol

34756-0050 34756-0250
TS34756-0050EA 72 USD
TS34756-0050 TS34756-0250
3-Mercapto-1-hexanol 98%
3-Mercapto-1-hexanol
Formula: C₆H₁₄OS
MW: 134.24 g/mol
Boiling Pt: 45 °C (0.001 mmHg)
Density: 0.97 g/cm³
Flash Pt: >100 °C
MDL Number: MFCD00792515
CAS Number: 51755-83-0

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4790 to 1.4810 (20 °C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR