Ethylene glycol monostearate
Supplier: Thermo Scientific Chemicals
34695-1000
34695-2500
TS34695-1000EA
25
USD
TS34695-1000
TS34695-2500
Ethylene glycol monostearate
Ethylene glycol monostearate
Formula:
C₂₀H₄₀O₃ Boiling Pt: 149 °C Melting Pt: 55…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051465 CAS Number: 111-60-4 |
Specification Test Results
Appearance | White to light yellow Flakes |
Infrared spectrum | Conforms |
Melting point | 55 to 60 °C |
Additional info | compound is a mixture of various chain-length esters and di(ethylene glycol) ester |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...