trans-4-Aminocyclohexanol 97%

Supplier: Thermo Scientific Chemicals
Danger

34668-0050 34668-0250 34668-1000
TS34668-0050EA 41 USD
TS34668-0050 TS34668-0250 TS34668-1000
trans-4-Aminocyclohexanol 97%
trans-4-Aminocyclohexanol
Formula: C₆H₁₃NO
Boiling Pt: 127 °C (14 mmHg)
Melting Pt: 108…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00067698
CAS Number: 27489-62-9
EINECS: 248-492-8
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Appearance White to cream to light brown Powder or crystals
Infrared spectrum Conforms
Melting point 108 to 113 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR