Bromoacetaldehyde diethyl acetal 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Bromo-1,1-diethoxyethane

34652-1000 34652-5000
TS34652-1000EA 51 USD
TS34652-1000 TS34652-5000
Bromoacetaldehyde diethyl acetal 97%
Bromoacetaldehyde diethyl acetal
Formula: BrH₂CCH(OCH₂CH₃)₂
Boiling Pt: 66…67 °C (18 mmHg)
Density: 1.28 g/cm³
Flash Pt: 61 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000214
CAS Number: 2032-35-1
EINECS: 217-989-1
UN: 2810
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4380 to 1.4400 (20 °C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR