2,4-Dichloro-6,7-dimethoxyquinazoline 97%

Supplier: Thermo Scientific Chemicals
Warning

34255-0050 34255-0250
TS34255-0050EA 67 USD
TS34255-0050 TS34255-0250
2,4-Dichloro-6,7-dimethoxyquinazoline 97%
2,4-Dichloro-6,7-dimethoxyquinazoline
Formula: C₁₀H₈Cl₂N₂O₂
Melting Pt: 172…177 °C
Storage Temperature: Ambient
MDL Number: MFCD00051733
CAS Number: 27631-29-4

Order Now

Specification Test Results

Appearance White to yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 172 to 179 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR