Methyl-2-oxocyclopentanecarboxylate 96%

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl 2-cyclopentanonecarboxylate

34209-0050 34209-0250 34209-1000
TS34209-0050EA 25 USD
TS34209-0050 TS34209-0250 TS34209-1000
Methyl-2-oxocyclopentanecarboxylate 96%
Methyl-2-oxocyclopentanecarboxylate
Formula: C₇H₁₀O₃
Boiling Pt: 102…104 °C (11 mmHg)
Density: 1.054 g/cm³
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00001411
CAS Number: 10472-24-9
EINECS: 233-962-7

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4550 to 1.4570 (20 °C, 589 nm)
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR