tert-Butyl methyl malonate 95%

Supplier: Thermo Scientific Chemicals

Synonyms: tert.-Butyl methyl malonate

33939-0050 33939-0250
TS33939-0010EA 33.07 USD
TS33939-0010 TS33939-0050 TS33939-0250
tert-Butyl methyl malonate 95%
tert-Butyl methyl malonate
Formula: C₈H₁₄O₄
MW: 174.19 g/mol
Boiling Pt: 80 °C (11 mmHg)
Melting Pt: –10 °C
Density: 1.03 g/cm³
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00042856
CAS Number: 42726-73-8
EINECS: 255-919-1

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid (May discolor to yellow on storage)
Infrared spectrum Conforms
Refractive index 1.4140 to 1.4170 (20 °C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR