Hexadecyl palmitate 98%

Supplier: Thermo Scientific Chemicals

33846-0050 33846-0250 33846-1000
TS33846-0050EA 51 USD
TS33846-0050 TS33846-0250 TS33846-1000
Hexadecyl palmitate 98%
Hexadecyl palmitate
Formula: C₃₂H₆₄O₂
Boiling Pt: 360 °C (760 mmHg)
Melting Pt: 52…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00053739
CAS Number: 540-10-3

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Melting point 52 to 56 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR