4-Isopropylbenzaldehyde 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Cuminaldehyde

33488-0050 33488-1000 33488-5000
TS33488-0050EA 36 USD
TS33488-0050 TS33488-1000 TS33488-5000
4-Isopropylbenzaldehyde 98%
4-Isopropylbenzaldehyde
Formula: C₁₀H₁₂O
MW: 148.2 g/mol
MDL Number: MFCD00006953
CAS Number: 122-03-2

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5290 to 1.5320 (20 °C, 589 nm)
Density (20°C) 0.975 to 0.983 g/ml
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR