(S)-(+)-2-Methoxy-1-propanol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-(+)-2-Methoxypropanol

33185-0010 33185-0025 33185-0100
TS33185-0010EA 167 USD
TS33185-0010 TS33185-0025 TS33185-0100
(S)-(+)-2-Methoxy-1-propanol 97%
(S)-(+)-2-Methoxy-1-propanol
Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Density: 0.938 g/cm³
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD01632593
CAS Number: 116422-39-0
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4060 to 1.4090 (20 °C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥96.0 %
Specific optical rotation +38° to +44° (20°C, 589 nm) (c=1.66, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR