(S)-(+)-1-Methoxy-2-propanol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-1-Methoxypropan-2-ol, (S)-(+)-Propylene glycol 1-monomethyl ether

33179-0050 33179-0250
TS33179-0050EA 322 USD
TS33179-0050 TS33179-0250
(S)-(+)-1-Methoxy-2-propanol 99%
(S)-(+)-1-Methoxy-2-propanol
Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Melting Pt: –97 °C
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD01632588
CAS Number: 26550-55-0
UN: 3092
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4020 to 1.4040 (20 °C, 589 nm)
GC ≥98.5 %
Specific optical rotation +20° to +24° (25°C, 589 nm) (c=10, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR