(±)-2-Decanol 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (±)-Methyl octyl carbinol, (±)-2-Hydroxy decane, 2-Decanol, (±)-Octyl methyl carbinol

33160-0050 33160-0250
TS33160-0050EA 30 USD
TS33160-0050 TS33160-0250
(±)-2-Decanol 98%
(±)-2-Decanol
Formula: C₁₀H₂₂O
MW: 158.28 g/mol
Boiling Pt: 211
Melting Pt: -5
Density: 0.82 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00004594
CAS Number: 1120-06-5
EINECS: 214-296-6

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4330 to 1.4350 (20 °C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR