1-(6-Methoxy-2-naphthyl)ethanol 98%

Supplier: Thermo Scientific Chemicals

Synonyms: DL-6-Methoxy-α-methyl-2-naphthalenemethanol, 1-(6-Methoxynaphthalen-2-yl)ethanol

33150-0050 33150-0250
TS33150-0050EA 77 USD
TS33150-0050 TS33150-0250
1-(6-Methoxy-2-naphthyl)ethanol 98%
1-(6-Methoxy-2-naphthyl)ethanol
Formula: C₁₃H₁₄O₂
MW: 202.25 g/mol
Melting Pt: 111…114 °C
Storage Temperature: Ambient
MDL Number: MFCD01632581
CAS Number: 77301-42-9

Order Now

Specification Test Results

Appearance White to yellow to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 111 to 114 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR