2'-Hydroxy-5'-methoxyacetophenone 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2'-Hydroxy-5'-Methoxyacetophenone, 2′-Hydroxy-5′-methoxyacetophenone

33124-0050 33124-0250
TS33124-0050EA 42 USD
TS33124-0050 TS33124-0250
2'-Hydroxy-5'-methoxyacetophenone 99%
2'-Hydroxy-5'-methoxyacetophenone
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 281 °C (1013 hPa)
Melting Pt: 48…51 °C
Flash Pt: >110 °C
MDL Number: MFCD00008731
CAS Number: 705-15-7
EINECS: 211-882-3

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 48 to 51 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR