3-Butyne-2-one 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: But-3-yn-2-one

33044-0025 33044-0100
TS33044-0025EA 119 USD
TS33044-0025 TS33044-0100
3-Butyne-2-one 96%
3-Butyne-2-one
Formula: C₄H₄O
MW: 68.08 g/mol
Boiling Pt: 85 °C (760 mmHg)
Density: 0.87 g/cm³
Flash Pt: –1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008775
CAS Number: 1423-60-5
EINECS: 215-834-2
UN: 1992
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear yellow to orange-brown Liquid
Infrared spectrum Conforms
Refractive index 1.4045 to 1.4095 (20 °C, 589 nm)
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR